siRNA Details
Gene Symbol: RABL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306160.1 | NP_001293089.1 | 5, 6 | 459 | ||
NM_007082.4 | NP_009013.1 | 6, 7 | 631 | |||
NM_013412.3 | NP_038198.1 | 6, 7 | 631 | |||
XM_017003208.1 | XP_016858697.1 | 6, 7 | 625 | |||
XM_017003209.1 | XP_016858698.1 | 6, 7 | 609 | |||
XM_017003212.1 | XP_016858701.1 | 5, 6 | 1351 | |||
XM_017003220.1 | XP_016858709.1 | 5, 6 | 1351 | |||
XM_017003221.1 | XP_016858710.1 | 6, 7 | 627 | |||
XM_017003222.1 | XP_016858711.1 | 7, 8 | 782 | |||
GenBank mRNA | AF095350.1 | - | 6, 7 | 559 | ||
AF095351.1 | - | 6, 7 | 559 | |||
CN342140.1 | - | 5, 6 | 463 |
Gene Symbol: RABL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130922.1 | NP_001124394.1 | 6, 7 | 622 | ||
NM_001130923.1 | NP_001124395.1 | 7, 8 | 777 | |||
NM_007081.2 | NP_009012.1 | 6, 7 | 618 | |||
XM_005261899.2 | XP_005261956.1 | 7, 8 | 874 | |||
XM_011530675.1 | XP_011528977.1 | 7, 8 | 874 | |||
XM_017028547.1 | XP_016884036.1 | 7, 8 | 1064 | |||
XM_017028548.1 | XP_016884037.1 | 6, 7 | 621 | |||
XM_017028549.1 | XP_016884038.1 | 7, 8 | 910 | |||
XM_017028551.1 | XP_016884040.1 | 5, 6 | 795 | |||
XM_017028556.1 | XP_016884045.1 | 7, 8 | 1064 | |||
XM_017028557.1 | XP_016884046.1 | 7, 8 | 910 | |||
XM_017028558.1 | XP_016884047.1 | 7, 8 | 748 | |||
GenBank mRNA | AF095352.1 | - | 6, 7 | 559 | ||
BG177165.1 | - | 619 | ||||
BX432293.2 | - | 6, 7 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
