siRNA Details
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291505.1 | NP_001278434.1 | 3, 4 | 702 | ||
NM_001316325.1 | NP_001303254.1 | 5, 6 | 925 | |||
NM_006241.7 | NP_006232.1 | 4, 5 | 780 | |||
GenBank mRNA | AJ133812.1 | - | 4, 5 | 407 | ||
AY063767.1 | - | 4, 5 | 406 | |||
BC007655.2 | - | 4, 5 | 629 | 3346573 | ||
X78873.1 | - | 4, 5 | 407 | |||
Z29646.1 | - | 4, 5 | 404 |
Gene Symbol: PPP1R2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF275684.1 | - | 1 | 419 |
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 429 | |||
GenBank mRNA | BC066922.1 | - | 1 | 429 | ||
JF438008.1 | - | 1 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
