siRNA Details
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291504.1 | NP_001278433.1 | 5 | 833 | ||
NM_001291505.1 | NP_001278434.1 | 4 | 752 | |||
NM_001316325.1 | NP_001303254.1 | 6 | 975 | |||
NM_006241.7 | NP_006232.1 | 5 | 830 | |||
NR_133014.1 | 4 | 735 | ||||
XM_006713682.3 | XP_006713745.1 | 5 | 779 | |||
GenBank mRNA | AJ133812.1 | - | 5 | 457 | ||
AY063767.1 | - | 5 | 456 | |||
BC007655.2 | - | 5 | 679 | 3346573 | ||
BI549862.1 | - | 4 | 654 | |||
X78873.1 | - | 5 | 457 | |||
Z29646.1 | - | 5 | 454 |
Gene Symbol: PPP1R2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF275684.1 | - | 1 | 469 |
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 479 | |||
GenBank mRNA | BC066922.1 | - | 1 | 479 | ||
JF438008.1 | - | 1 | 531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
