siRNA Details
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 24 | 5162 | ||
XM_005254137.3 | XP_005254194.1 | 25 | 5334 | |||
XM_011521166.2 | XP_011519468.1 | 14 | 3627 | |||
GenBank mRNA | AL110226.1 | - | 9 | 2556 | ||
AL359934.1 | - | 6 | 1159 | |||
AY327122.1 | - | 24 | 4929 | |||
BC061631.1 | - | 24 | 5142 | 6650221 |
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045529.3 | 24 | 4681 | |||
GenBank mRNA | AK131095.1 | - | 4 | 5321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
