siRNA Details
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 19 | 4019 | ||
XM_005254137.3 | XP_005254194.1 | 19 | 4020 | |||
XM_011521166.2 | XP_011519468.1 | 8 | 2313 | |||
GenBank mRNA | AL110226.1 | - | 4 | 1413 | ||
AL359934.1 | - | 1 | 16 | |||
AY327122.1 | - | 19 | 3786 | |||
BC061631.1 | - | 19 | 3999 | 6650221 |
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045529.3 | 19 | 3533 | |||
GenBank mRNA | AK131095.1 | - | 1 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
