siRNA Details
Gene Symbol: NDUFV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021074.4 | NP_066552.2 | 3 | 284 | ||
XM_017025782.1 | XP_016881271.1 | 3 | 474 | |||
GenBank mRNA | AK291865.1 | - | 3 | 180 | ||
AK312129.1 | - | 3 | 202 | |||
BC001632.1 | - | 3 | 194 | 3536345 | ||
BC017487.1 | - | 3 | 192 | 3537815 | ||
BC071689.1 | - | 3 | 174 | 4372795 | ||
BG742948.1 | - | 3 | 284 | |||
CR456928.1 | - | 3 | 170 | |||
M22538.1 | - | 3 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
