siRNA Details
Gene Symbol: ING5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032329.4 | NP_115705.2 | 8 | 961 | ||
XM_017005097.1 | XP_016860586.1 | 8 | 2029 | |||
XM_017005098.1 | XP_016860587.1 | 9 | 2078 | |||
GenBank mRNA | AF189286.1 | - | 8 | 938 | ||
AK074422.1 | - | 8 | 961 | |||
AK289958.1 | - | 8 | 962 | |||
BC071899.1 | - | 8 | 952 | 6111003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
