siRNA Details
Gene Symbol: ING5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032329.4 | NP_115705.2 | 8 | 1496 | ||
XM_017005097.1 | XP_016860586.1 | 8 | 2564 | |||
XM_017005098.1 | XP_016860587.1 | 9 | 2613 | |||
GenBank mRNA | AI701412.1 | - | 1 | 395 | ||
AK074422.1 | - | 8 | 1496 | |||
BC071899.1 | - | 8 | 1487 | 6111003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
