siRNA Details
Gene Symbol: ZNF736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 1377 | ||
GenBank mRNA | AK301806.1 | - | 4 | 1221 |
Gene Symbol: ZNF727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012225.1 | XP_016867714.1 | 3 | 2403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
