siRNA Details
Gene Symbol: TBC1D3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001417.6 | NP_001001417.6 | 6 | 491 | ||
XM_005257980.4 | XP_005258037.2 | 6 | 621 | |||
GenBank mRNA | AF540953.1 | - | 5 | 345 | ||
AK292257.1 | - | 6 | 462 |
Gene Symbol: TBC1D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK313329.1 | - | 6 | 485 |
Gene Symbol: TBC1D3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047739.1 | - | 7 | 656 |
Gene Symbol: TBC1D3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032258.4 | NP_115634.3 | 6 | 459 | ||
GenBank mRNA | AK225985.1 | - | 6 | 450 | ||
AK292344.1 | - | 6 | 502 | |||
BC075809.1 | - | 6 | 491 | 5170064 |
Gene Symbol: TBC1D3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291462.1 | NP_001278391.1 | 6 | 445 | ||
XM_005276914.3 | XP_005276971.1 | 6 | 2270 | |||
GenBank mRNA | AK292308.1 | - | 6 | 495 | ||
AK301823.1 | - | 6 | 444 | |||
BC071680.1 | - | 6 | 445 | 5165385 |
Gene Symbol: TBC1D3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291464.1 | NP_001278393.1 | 6 | 445 | ||
XM_006722235.2 | XP_006722298.1 | 5 | 2723 | |||
XM_006722236.1 | XP_006722299.1 | 6 | 1295 | |||
XM_006722237.2 | XP_006722300.1 | 5 | 2540 | |||
XM_006722239.2 | XP_006722302.1 | 5 | 2540 | |||
XM_011524172.1 | XP_011522474.1 | 5 | 2723 | |||
XM_011524173.1 | XP_011522475.1 | 5 | 2723 | |||
XM_011524174.1 | XP_011522476.1 | 5 | 2723 | |||
GenBank mRNA | DC397859.1 | - | 6 | 478 |
Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB024853.1 | - | 6 | 457 |
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 6 | 457 | |||
GenBank mRNA | AK301746.1 | - | 6 | 458 | ||
BC058890.1 | - | 6 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
