siRNA Details
Gene Symbol: KRT19P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036685.1 | 1 | 15 | |||
GenBank mRNA | AB041269.1 | - | 1 | 15 | ||
AB041270.1 | - | 1 | 15 |
Gene Symbol: MIR492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030171.1 | 1 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
