siRNA Details
Gene Symbol: AKR7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040288.1 | 7 | 1029 | |||
NR_040289.1 | 5 | 832 | ||||
GenBank mRNA | AJ278012.1 | - | 6 | 815 | ||
BC035351.1 | - | 5 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
