siRNA Details
Gene Symbol: PCYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184917.2 | NP_001171846.1 | 3 | 345 | ||
NM_001256434.2 | NP_001243363.1 | 3 | 343 | |||
NM_001256435.2 | NP_001243364.1 | 3 | 629 | |||
NM_001282203.1 | NP_001269132.1 | 3 | 757 | |||
NM_001282204.1 | NP_001269133.1 | 3 | 473 | |||
NM_002861.4 | NP_002852.1 | 3 | 345 | |||
XM_005256386.3 | XP_005256443.1 | 3 | 495 | |||
XM_005256387.3 | XP_005256444.1 | 3 | 415 | |||
XM_006722287.3 | XP_006722350.1 | 3 | 270 | |||
XM_017024910.1 | XP_016880399.1 | 3 | 270 | |||
XM_017024911.1 | XP_016880400.1 | 3 | 415 | |||
XM_017024912.1 | XP_016880401.1 | 3 | 246 | |||
GenBank mRNA | AK093328.1 | - | 3 | 693 | ||
AK297996.1 | - | 3 | 406 | |||
AK301723.1 | - | 3 | 507 | |||
AK315777.1 | - | 3 | 267 | |||
AK316385.1 | - | 3 | 166 | |||
AY251539.1 | - | 3 | 236 | |||
BC000351.2 | - | 3 | 253 | 2961418 | ||
BC010075.2 | - | 3 | 296 | 4303545 | ||
CR456779.1 | - | 3 | 223 | |||
D84307.1 | - | 3 | 289 | |||
HY042603.1 | - | 3 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
