siRNA Details
Gene Symbol: PCYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184917.2 | NP_001171846.1 | 5 | 556 | ||
NM_001256433.2 | NP_001243362.1 | 4 | 394 | |||
NM_001256434.2 | NP_001243363.1 | 5 | 554 | |||
NM_001256435.2 | NP_001243364.1 | 5 | 840 | |||
NM_001282203.1 | NP_001269132.1 | 5 | 968 | |||
NM_001282204.1 | NP_001269133.1 | 5 | 684 | |||
NM_002861.4 | NP_002852.1 | 5 | 556 | |||
XM_005256386.3 | XP_005256443.1 | 5 | 706 | |||
XM_005256387.3 | XP_005256444.1 | 5 | 626 | |||
XM_006722287.3 | XP_006722350.1 | 5 | 481 | |||
XM_017024910.1 | XP_016880399.1 | 5 | 481 | |||
XM_017024911.1 | XP_016880400.1 | 5 | 626 | |||
XM_017024912.1 | XP_016880401.1 | 5 | 457 | |||
GenBank mRNA | AK093328.1 | - | 5 | 904 | ||
AK297996.1 | - | 5 | 617 | |||
AK301723.1 | - | 5 | 718 | |||
AK315777.1 | - | 5 | 478 | |||
AK316385.1 | - | 5 | 377 | |||
AY251539.1 | - | 5 | 447 | |||
BC000351.2 | - | 5 | 464 | 2961418 | ||
BC010075.2 | - | 5 | 507 | 4303545 | ||
BX459568.2 | - | 5 | 542 | |||
CR456779.1 | - | 5 | 434 | |||
D84307.1 | - | 5 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
