siRNA Details
Gene Symbol: PCYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184917.2 | NP_001171846.1 | 9, 10 | 932 | ||
NM_001256433.2 | NP_001243362.1 | 7, 8 | 716 | |||
NM_001256434.2 | NP_001243363.1 | 9, 10 | 930 | |||
NM_001256435.2 | NP_001243364.1 | 8, 9 | 1162 | |||
NM_001282203.1 | NP_001269132.1 | 8, 9 | 1290 | |||
NM_001282204.1 | NP_001269133.1 | 8, 9 | 1006 | |||
NM_002861.4 | NP_002852.1 | 8, 9 | 878 | |||
XM_005256386.3 | XP_005256443.1 | 9, 10 | 1082 | |||
XM_005256387.3 | XP_005256444.1 | 9, 10 | 1002 | |||
XM_006722287.3 | XP_006722350.1 | 9, 10 | 857 | |||
XM_017024910.1 | XP_016880399.1 | 8, 9 | 803 | |||
XM_017024911.1 | XP_016880400.1 | 8, 9 | 948 | |||
XM_017024912.1 | XP_016880401.1 | 9, 10 | 1054 | |||
GenBank mRNA | AK093328.1 | - | 8, 9 | 1226 | ||
AK297996.1 | - | 8, 9 | 939 | |||
AK301723.1 | - | 8, 9 | 1040 | |||
AK315777.1 | - | 9, 10 | 854 | |||
AK316385.1 | - | 8, 9 | 699 | |||
AY251539.1 | - | 9, 10 | 823 | |||
BC000351.2 | - | 8, 9 | 786 | 2961418 | ||
BC010075.2 | - | 8, 9 | 829 | 4303545 | ||
BX419403.2 | - | 7, 8 | 656 | |||
CR456779.1 | - | 8, 9 | 756 | |||
D84307.1 | - | 8, 9 | 822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
