siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052588.1 | - | 2 | 139 | ||
BF984173.1 | - | 2 | 135 | |||
D38436.1 | - | 2 | 117 | |||
D38439.1 | - | 1 | 48 | |||
D38501.1 | - | 1 | 48 | |||
D38502.1 | - | 2 | 117 | |||
DW407867.1 | - | 2 | 153 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DR007973.1 | - | 2 | 316 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 2 | 91 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 2 | 527 | |||
GenBank mRNA | AB017004.1 | - | 2 | 527 | ||
BC067809.1 | - | 2 | 159 | |||
BC110395.1 | - | 2 | 135 | |||
D38435.1 | - | 2 | 130 | |||
D38499.1 | - | 2 | 130 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 2 | 87 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 2 | 111 | |||
GenBank mRNA | DA215155.1 | - | 2 | 98 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 2 | 124 | |||
NR_046297.1 | 2 | 124 | ||||
NR_046299.1 | 2 | 124 | ||||
GenBank mRNA | BC029368.1 | - | 2 | 101 | ||
BC029419.1 | - | 2 | 89 | |||
CB141754.1 | - | 2 | 89 | |||
D38438.1 | - | 2 | 95 | |||
D38500.1 | - | 2 | 95 | |||
DA885243.1 | - | 2 | 124 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 3 | 289 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 2 | 152 | |||
GenBank mRNA | AB017006.1 | - | 2 | 247 | ||
DB472687.1 | - | 2 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
