siRNA Details
Gene Symbol: D2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287249.1 | NP_001274178.1 | 7, 8 | 961 | ||
NM_152783.4 | NP_689996.4 | 8, 9 | 1345 | |||
XM_011511734.1 | XP_011510036.1 | 9, 10 | 1374 | |||
XM_011511735.1 | XP_011510037.1 | 9, 10 | 1374 | |||
XM_011511736.1 | XP_011510038.1 | 8, 9 | 1296 | |||
XM_011511737.2 | XP_011510039.1 | 9, 10 | 1374 | |||
XM_011511754.1 | XP_011510056.1 | 6, 7 | 796 | |||
XM_017004828.1 | XP_016860317.1 | 8, 9 | 1296 | |||
XM_017004831.1 | XP_016860320.1 | 7, 8 | 1167 | |||
GenBank mRNA | AK094149.1 | - | 8, 9 | 938 | ||
AK315188.1 | - | 2, 3 | 461 | |||
BC031817.1 | - | 2, 3 | 335 | |||
BC036604.2 | - | 8, 9 | 1284 | 5274813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
