siRNA Details
Gene Symbol: D2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287249.1 | NP_001274178.1 | 7 | 875 | ||
NM_152783.4 | NP_689996.4 | 8 | 1259 | |||
XM_011511734.1 | XP_011510036.1 | 9 | 1288 | |||
XM_011511735.1 | XP_011510037.1 | 9 | 1288 | |||
XM_011511736.1 | XP_011510038.1 | 8 | 1210 | |||
XM_011511737.2 | XP_011510039.1 | 9 | 1288 | |||
XM_011511743.1 | XP_011510045.1 | 10 | 1425 | |||
XM_011511744.1 | XP_011510046.1 | 10 | 1425 | |||
XM_011511745.2 | XP_011510047.1 | 9 | 1288 | |||
XM_011511750.2 | XP_011510052.1 | 9 | 1288 | |||
XM_011511754.1 | XP_011510056.1 | 6 | 710 | |||
XM_011511757.2 | XP_011510059.1 | 10 | 1321 | |||
XM_017004828.1 | XP_016860317.1 | 8 | 1210 | |||
XM_017004829.1 | XP_016860318.1 | 10 | 1425 | |||
XM_017004830.1 | XP_016860319.1 | 9 | 1288 | |||
XM_017004831.1 | XP_016860320.1 | 7 | 1081 | |||
GenBank mRNA | AK055846.1 | - | 1 | 37 | ||
AK094149.1 | - | 8 | 852 | |||
AK124189.1 | - | 1 | 726 | |||
AK304773.1 | - | 10 | 1387 | |||
AK315188.1 | - | 2 | 375 | |||
BC031817.1 | - | 2 | 249 | |||
BC036604.2 | - | 8 | 1198 | 5274813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
