siRNA Details
Gene Symbol: USP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032582.3 | NP_115971.2 | 32 | 4432 | ||
XM_011525371.1 | XP_011523673.1 | 32 | 4355 | |||
XM_011525372.1 | XP_011523674.1 | 32 | 4313 | |||
XM_011525373.1 | XP_011523675.1 | 32 | 4383 | |||
XM_011525374.1 | XP_011523676.1 | 32 | 4200 | |||
XM_011525375.1 | XP_011523677.1 | 32 | 4173 | |||
XM_011525376.1 | XP_011523678.1 | 32 | 4158 | |||
XM_011525378.1 | XP_011523680.1 | 31 | 4065 | |||
XM_011525379.2 | XP_011523681.1 | 28 | 3688 | |||
XM_017025233.1 | XP_016880722.1 | 31 | 4097 | |||
XM_017025234.1 | XP_016880723.1 | 24 | 3235 | |||
GenBank mRNA | AF350251.1 | - | 24 | 3185 | ||
AF533230.1 | - | 32 | 4318 | |||
BC070497.1 | - | 3 | 436 | |||
BC094680.1 | - | 3 | 501 |
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 6 | 1163 | |||
GenBank mRNA | AK302945.1 | - | 2 | 268 | ||
AK316466.1 | - | 4 | 881 | |||
AY363305.1 | - | 2 | 545 | |||
BC039345.1 | - | 6 | 920 |
Gene Symbol: USP32P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003554.1 | 3 | 2083 | |||
GenBank mRNA | AJ012755.1 | - | 3 | 1637 | ||
AK308293.1 | - | 6 | 795 |
Gene Symbol: USP32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310665.1 | - | 3 | 367 |
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 36 | 5770 | ||
NM_004505.3 | NP_004496.2 | 28 | 5239 | |||
XM_011524050.1 | XP_011522352.1 | 32 | 5988 | |||
XM_011524051.2 | XP_011522353.1 | 34 | 5083 | |||
XM_011524052.2 | XP_011522354.1 | 33 | 5197 | |||
XM_011524053.2 | XP_011522355.1 | 34 | 5110 | |||
XM_011524054.2 | XP_011522356.1 | 34 | 5077 | |||
XM_011524055.2 | XP_011522357.1 | 32 | 5004 | |||
XM_011524056.2 | XP_011522358.1 | 29 | 4957 | |||
XM_011524057.2 | XP_011522359.1 | 32 | 5120 | |||
XM_017025290.1 | XP_016880779.1 | 31 | 5826 | |||
GenBank mRNA | AY143550.1 | - | 28 | 5239 | ||
BX647719.1 | - | 36 | 5772 | |||
X63546.1 | - | 27 | 5124 | |||
X63547.1 | - | 29 | 5447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
