siRNA Details
Gene Symbol: USP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032582.3 | NP_115971.2 | 32 | 4535 | ||
XM_011525371.1 | XP_011523673.1 | 32 | 4458 | |||
XM_011525372.1 | XP_011523674.1 | 32 | 4416 | |||
XM_011525373.1 | XP_011523675.1 | 32 | 4486 | |||
XM_011525374.1 | XP_011523676.1 | 32 | 4303 | |||
XM_011525375.1 | XP_011523677.1 | 32 | 4276 | |||
XM_011525376.1 | XP_011523678.1 | 32 | 4261 | |||
XM_011525378.1 | XP_011523680.1 | 31 | 4168 | |||
XM_011525379.2 | XP_011523681.1 | 28 | 3791 | |||
XM_017025233.1 | XP_016880722.1 | 31 | 4200 | |||
XM_017025234.1 | XP_016880723.1 | 24 | 3338 | |||
GenBank mRNA | AF350251.1 | - | 24 | 3288 | ||
AF533230.1 | - | 32 | 4421 | |||
BC070497.1 | - | 3 | 539 | |||
BC094680.1 | - | 3 | 604 |
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 6 | 1266 | |||
GenBank mRNA | AK302945.1 | - | 2 | 371 | ||
AK316466.1 | - | 4 | 984 | |||
AY363305.1 | - | 2 | 648 | |||
BC039345.1 | - | 6 | 1023 |
Gene Symbol: USP32P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003554.1 | 3 | 2186 | |||
GenBank mRNA | AJ012755.1 | - | 3 | 1740 | ||
AK124785.1 | - | 8 | 2106 | |||
AK308293.1 | - | 6 | 898 |
Gene Symbol: USP32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310665.1 | - | 3 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
