siRNA Details
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 36 | 5896 | ||
NM_004505.3 | NP_004496.2 | 28 | 5365 | |||
XM_011524050.1 | XP_011522352.1 | 32 | 6114 | |||
XM_011524051.2 | XP_011522353.1 | 34 | 5209 | |||
XM_011524052.2 | XP_011522354.1 | 33 | 5323 | |||
XM_011524053.2 | XP_011522355.1 | 34 | 5236 | |||
XM_011524054.2 | XP_011522356.1 | 34 | 5203 | |||
XM_011524055.2 | XP_011522357.1 | 32 | 5130 | |||
XM_011524056.2 | XP_011522358.1 | 29 | 5083 | |||
XM_011524057.2 | XP_011522359.1 | 32 | 5246 | |||
XM_017025290.1 | XP_016880779.1 | 31 | 5952 | |||
GenBank mRNA | AY143550.1 | - | 28 | 5365 | ||
BX647719.1 | - | 36 | 5898 | |||
X63546.1 | - | 27 | 5250 | |||
X63547.1 | - | 29 | 5573 |
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 6 | 1289 | |||
GenBank mRNA | AK302945.1 | - | 2 | 394 | ||
AK316466.1 | - | 4 | 1007 | |||
AY363305.1 | - | 2 | 671 |
Gene Symbol: USP32P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003554.1 | 3 | 2209 | |||
GenBank mRNA | AJ012755.1 | - | 3 | 1763 | ||
AK124785.1 | - | 8 | 2129 | |||
AK308293.1 | - | 6 | 921 |
Gene Symbol: USP32P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310665.1 | - | 3 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
