siRNA Details
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004562.2 | NP_004553.2 | 2 | 259 | ||
NM_013987.2 | NP_054642.2 | 2 | 259 | |||
NM_013988.2 | NP_054643.2 | 2 | 259 | |||
XM_011535863.1 | XP_011534165.1 | 2 | 259 | |||
XM_017010908.1 | XP_016866397.1 | 2 | 1173 | |||
GenBank mRNA | AB009973.1 | - | 2 | 226 | ||
AB245403.1 | - | 2 | 217 | |||
AF381282.1 | - | 1 | 125 | |||
AF381284.1 | - | 1 | 125 | |||
AK292590.1 | - | 2 | 220 | |||
BC022014.2 | - | 2 | 223 | 4824892 | ||
DB023187.1 | - | 2 | 259 | |||
EF375726.1 | - | 1 | 125 | |||
GU345838.1 | - | 1 | 142 | |||
GU345839.1 | - | 2 | 142 | |||
GU345840.1 | - | 1 | 142 | |||
GU361466.1 | - | 1 | 142 | |||
GU361467.1 | - | 1 | 142 | |||
GU361468.1 | - | 1 | 142 | |||
GU361469.1 | - | 2 | 239 | |||
GU361470.1 | - | 2 | 240 | |||
KC357595.1 | - | 4 | 337 | |||
KJ534911.1 | - | 1 | 125 | |||
KJ534912.1 | - | 1 | 125 | |||
KU178240.1 | - | 1 | 125 | |||
KU178241.1 | - | 1 | 125 | |||
KU178242.1 | - | 1 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
