siRNA Details
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004562.2 | NP_004553.2 | 6 | 787 | ||
NM_013987.2 | NP_054642.2 | 5 | 703 | |||
NM_013988.2 | NP_054643.2 | 3 | 340 | |||
XM_011535863.1 | XP_011534165.1 | 6 | 784 | |||
XM_017010908.1 | XP_016866397.1 | 6 | 1701 | |||
XM_017010909.1 | XP_016866398.1 | 5 | 581 | |||
GenBank mRNA | AB009973.1 | - | 6 | 754 | ||
AB245403.1 | - | 6 | 745 | |||
AF381282.1 | - | 4 | 412 | |||
AF381283.1 | - | 4 | 489 | |||
AF381284.1 | - | 4 | 531 | |||
AK292590.1 | - | 6 | 748 | |||
BC022014.2 | - | 6 | 751 | 4824892 | ||
EF375726.1 | - | 5 | 653 | |||
GU345837.1 | - | 5 | 506 | |||
GU345838.1 | - | 4 | 548 | |||
GU345839.1 | - | 5 | 586 | |||
GU345840.1 | - | 5 | 670 | |||
GU357501.1 | - | 3 | 143 | |||
GU357502.1 | - | 2 | 143 | |||
GU361466.1 | - | 4 | 548 | |||
GU361467.1 | - | 4 | 586 | |||
GU361468.1 | - | 4 | 429 | |||
GU361469.1 | - | 6 | 767 | |||
GU361470.1 | - | 6 | 768 | |||
KC357595.1 | - | 8 | 865 | |||
KC774171.1 | - | 4 | 520 | |||
KJ534911.1 | - | 5 | 653 | |||
KJ534912.1 | - | 5 | 653 | |||
KU178240.1 | - | 5 | 653 | |||
KU178241.1 | - | 5 | 653 | |||
KU178242.1 | - | 5 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
