siRNA Details
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004562.2 | NP_004553.2 | 7 | 926 | ||
NM_013987.2 | NP_054642.2 | 6 | 842 | |||
NM_013988.2 | NP_054643.2 | 4 | 479 | |||
XM_011535863.1 | XP_011534165.1 | 7 | 923 | |||
XM_017010908.1 | XP_016866397.1 | 7 | 1840 | |||
XM_017010909.1 | XP_016866398.1 | 6 | 720 | |||
GenBank mRNA | AB009973.1 | - | 7 | 893 | ||
AB245403.1 | - | 8 | 884 | |||
AF381282.1 | - | 5 | 551 | |||
AF381283.1 | - | 5 | 628 | |||
AF381284.1 | - | 5 | 670 | |||
AK292590.1 | - | 7 | 887 | |||
AK294684.1 | - | 2 | 132 | |||
BC022014.2 | - | 7 | 890 | 4824892 | ||
EF375726.1 | - | 6 | 792 | |||
GU345837.1 | - | 6 | 645 | |||
GU345838.1 | - | 5 | 687 | |||
GU345839.1 | - | 6 | 725 | |||
GU345840.1 | - | 6 | 809 | |||
GU357501.1 | - | 4 | 282 | |||
GU357502.1 | - | 3 | 282 | |||
GU361466.1 | - | 5 | 687 | |||
GU361467.1 | - | 5 | 725 | |||
GU361468.1 | - | 5 | 568 | |||
GU361469.1 | - | 7 | 906 | |||
GU361470.1 | - | 7 | 907 | |||
KC357595.1 | - | 9 | 1004 | |||
KC774171.1 | - | 5 | 659 | |||
KJ534911.1 | - | 6 | 792 | |||
KJ534912.1 | - | 6 | 792 | |||
KU178240.1 | - | 6 | 792 | |||
KU178241.1 | - | 6 | 792 | |||
KU178242.1 | - | 6 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
