siRNA Details
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640980.1 | - | 5 | 761 |
Gene Symbol: CDK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131531.1 | - | 1 | 326 |
Gene Symbol: CEP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303401.1 | NP_001290330.1 | 11 | 6350 | ||
NM_024548.3 | NP_078824.2 | 11 | 6527 | |||
XM_006713743.3 | XP_006713806.1 | 10 | 6918 | |||
XM_011513127.1 | XP_011511429.1 | 11 | 6484 | |||
XM_017007178.1 | XP_016862667.1 | 10 | 6743 | |||
GenBank mRNA | AL833269.1 | - | 11 | 6477 |
Gene Symbol: CYP2T1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092778.1 | - | 1 | 1591 | ||
AK097370.1 | - | 2 | 1454 | |||
BC037804.1 | - | 1 | 1599 | |||
BC082980.1 | - | 1 | 1556 |
Gene Symbol: CYP7B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004820.4 | NP_004811.1 | 6 | 6245 | ||
XM_017014002.1 | XP_016869491.1 | 7 | 24643 |
Gene Symbol: SLC2A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF248053.1 | - | 5 | 2403 |
Gene Symbol: USP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL110213.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
