siRNA Details
Gene Symbol: C21orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162495.2 | NP_001155967.2 | 4 | 3812 | ||
NM_001162496.2 | NP_001155968.2 | 2 | 3656 | |||
NM_019596.5 | NP_062542.5 | 3 | 3737 | |||
GenBank mRNA | BC004959.1 | - | 1 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
