siRNA Details
Gene Symbol: NEIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256552.1 | NP_001243481.1 | 2 | 855 | ||
NM_024608.3 | NP_078884.2 | 2 | 931 | |||
NR_046311.1 | 3 | 1054 | ||||
XM_005254659.4 | XP_005254716.1 | 2 | 791 | |||
XM_006720677.3 | XP_006720740.1 | 2 | 776 | |||
XM_006720678.3 | XP_006720741.1 | 2 | 806 | |||
XM_006720679.3 | XP_006720742.1 | 2 | 714 | |||
XM_006720680.1 | XP_006720743.1 | 3 | 946 | |||
XM_006720681.1 | XP_006720744.1 | 3 | 793 | |||
XM_011522001.2 | XP_011520303.2 | 2 | 696 | |||
XM_011522002.1 | XP_011520304.1 | 3 | 919 | |||
XM_011522003.2 | XP_011520305.1 | 4 | 975 | |||
XM_011522004.2 | XP_011520306.1 | 2 | 663 | |||
XM_011522005.1 | XP_011520307.1 | 2 | 850 | |||
XM_017022566.1 | XP_016878055.1 | 2 | 778 | |||
XM_017022568.1 | XP_016878057.1 | 2 | 813 | |||
GenBank mRNA | AB079068.1 | - | 1 | 425 | ||
AK026055.1 | - | 2 | 852 | |||
AK097008.1 | - | 3 | 1054 | |||
AK128372.1 | - | 3 | 526 | |||
BC010876.1 | - | 2 | 836 | 3879198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
