siRNA Details
Gene Symbol: NEIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256552.1 | NP_001243481.1 | 10 | 1734 | ||
NM_024608.3 | NP_078884.2 | 10 | 1810 | |||
NR_046311.1 | 11 | 1933 | ||||
XM_005254659.4 | XP_005254716.1 | 10 | 1670 | |||
XM_006720677.3 | XP_006720740.1 | 10 | 1655 | |||
XM_006720678.3 | XP_006720741.1 | 9 | 1565 | |||
XM_006720679.3 | XP_006720742.1 | 10 | 1593 | |||
XM_006720680.1 | XP_006720743.1 | 11 | 1825 | |||
XM_006720681.1 | XP_006720744.1 | 11 | 1672 | |||
XM_011522001.2 | XP_011520303.2 | 10 | 1575 | |||
XM_011522002.1 | XP_011520304.1 | 11 | 1798 | |||
XM_011522003.2 | XP_011520305.1 | 12 | 1854 | |||
XM_011522004.2 | XP_011520306.1 | 10 | 1542 | |||
XM_011522005.1 | XP_011520307.1 | 11 | 1884 | |||
XM_011522006.1 | XP_011520308.1 | 10 | 1808 | |||
XM_011522007.1 | XP_011520309.1 | 11 | 1904 | |||
XM_011522008.1 | XP_011520310.1 | 11 | 1877 | |||
XM_017022566.1 | XP_016878055.1 | 9 | 1537 | |||
XM_017022567.1 | XP_016878056.1 | 10 | 1568 | |||
GenBank mRNA | AK026055.1 | - | 11 | 1731 | ||
AK026216.1 | - | 4 | 1087 | |||
AK097008.1 | - | 11 | 1933 | |||
AK128372.1 | - | 8 | 2978 | |||
BC010876.1 | - | 10 | 1715 | 3879198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
