siRNA Details
Gene Symbol: KIF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015254.3 | NP_056069.2 | 6 | 524 | ||
XM_005273458.1 | XP_005273515.1 | 6 | 524 | |||
XM_011544457.1 | XP_011542759.1 | 6 | 524 | |||
XM_011544458.1 | XP_011542760.1 | 6 | 524 | |||
XM_011544459.2 | XP_011542761.1 | 6 | 509 | |||
XM_011544460.2 | XP_011542762.1 | 6 | 516 | |||
XM_011544461.1 | XP_011542763.1 | 5 | 511 | |||
XM_017013257.1 | XP_016868746.1 | 6 | 524 | |||
GenBank mRNA | AB014539.3 | - | 6 | 494 | ||
AF279865.1 | - | 6 | 502 | |||
AK307848.1 | - | 6 | 503 | |||
BC005977.1 | - | 6 | 486 | |||
BC051859.1 | - | 3 | 388 | |||
DA068190.1 | - | 6 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
