siRNA Details
Gene Symbol: HMGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005517.3 | NP_005508.1 | 4, 5 | 339 | ||
GenBank mRNA | AK026562.1 | - | 4, 5 | 181 | ||
BC000378.1 | - | 4, 5 | 242 | 2820349 | ||
BC003050.1 | - | 4, 5 | 232 | 3543809 | ||
BC003689.1 | - | 4, 5 | 203 | |||
BC007124.1 | - | 1 | 29 | |||
BC014644.1 | - | 4, 5 | 222 | 3355317 | ||
BC032140.2 | - | 4, 5 | 231 | 5021546 | ||
BC070297.1 | - | 2 | 190 | 4282630 | ||
BC071707.1 | - | 4, 5 | 232 | 3849426 | ||
BC072010.1 | - | 4, 5 | 226 | 6273507 | ||
BC072011.1 | - | 4, 5 | 222 | 6647282 | ||
BC075837.1 | - | 4, 5 | 327 | 6272425 | ||
BC081567.1 | - | 4, 5 | 225 | 4131303 | ||
BC110390.1 | - | 4, 5 | 235 | 5190365 | ||
CR542122.1 | - | 1 | 137 | |||
M12623.1 | - | 4, 5 | 225 |
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020810.3 | NP_065861.3 | 5 | 4415 | ||
XM_005267916.3 | XP_005267973.2 | 5 | 4445 | |||
GenBank mRNA | AB037814.1 | - | 4 | 4287 | ||
BC063551.1 | - | 1 | 470 | |||
BC136606.1 | - | 5 | 4330 | 9020596 | ||
BC136607.1 | - | 5 | 4330 | 9020597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
