siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB035332.1 | - | 1 | 289 | ||
AB035333.1 | - | 1 | 289 | |||
AB035334.1 | - | 1 | 289 | |||
AB035335.1 | - | 1 | 289 | |||
AB035338.1 | - | 4 | 1571 | |||
BX161462.1 | - | 6 | 1419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
