siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 5 | 1478 | |||
GenBank mRNA | AB035332.1 | - | 2 | 3257 | ||
AB035333.1 | - | 2 | 3257 | |||
AB035334.1 | - | 3 | 2289 | |||
AB035335.1 | - | 2 | 1834 | |||
AB035337.1 | - | 5 | 1480 | |||
AB035338.1 | - | 5 | 1675 | |||
AF195821.1 | - | 1 | 80 | |||
BC041075.1 | - | 2 | 1648 | 5923478 | ||
BX161462.1 | - | 7 | 1523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
