siRNA Details
Gene Symbol: STK39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013233.2 | NP_037365.2 | 3 | 545 | ||
XM_005246465.2 | XP_005246522.1 | 3 | 491 | |||
XM_017003813.1 | XP_016859302.1 | 3 | 491 | |||
XM_017003814.1 | XP_016859303.1 | 3 | 491 | |||
XM_017003815.1 | XP_016859304.1 | 3 | 1831 | |||
XM_017003816.1 | XP_016859305.1 | 3 | 491 | |||
GenBank mRNA | AF030403.1 | - | 3 | 637 | ||
AF099989.1 | - | 3 | 564 | |||
KJ534948.1 | - | 4 | 328 | |||
KJ534949.1 | - | 4 | 328 | |||
KJ534950.1 | - | 4 | 340 | |||
KJ534951.1 | - | 4 | 319 | |||
KJ535059.1 | - | 4 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
