siRNA Details
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 1 | 225 | ||
GenBank mRNA | AK291552.1 | - | 1 | 183 | ||
K03181.1 | - | 1 | 13 |
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 1 | 186 | ||
GenBank mRNA | BC006290.2 | - | 1 | 147 | 3960125 | |
BC051378.1 | - | 1 | 147 | 3956670 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 1 | 193 | ||
GenBank mRNA | BC030994.1 | - | 1 | 188 | 4605021 | |
BX118262.1 | - | 1 | 193 | |||
BX380841.2 | - | 1 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
