siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336879.1 | - | 1 | 273 | ||
AF336883.1 | - | 1 | 258 | |||
AF336884.1 | - | 1 | 318 | |||
AF336886.1 | - | 1 | 246 | |||
AF338190.1 | - | 1 | 291 | |||
AF338195.1 | - | 1 | 309 | |||
AF338198.1 | - | 1 | 309 | |||
AF469044.1 | - | 1 | 258 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
