siRNA Details
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 300 | ||
AF336879.1 | - | 1 | 324 | |||
AF336882.1 | - | 1 | 360 | |||
AF336883.1 | - | 1 | 309 | |||
AF336884.1 | - | 1 | 369 | |||
AF336886.1 | - | 1 | 297 | |||
AF338193.1 | - | 1 | 360 | |||
AF338195.1 | - | 1 | 360 | |||
AF338198.1 | - | 1 | 360 | |||
AF469044.1 | - | 1 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
