siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3853 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 2919 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3720 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3407 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 3982 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4246 | |||
NM_003879.5 | NP_003870.4 | 10 | 3825 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4720 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3689 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3768 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4655 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6672 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4528 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3866 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3491 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 153 | ||
AF336879.1 | - | 1 | 177 | |||
AF336883.1 | - | 1 | 162 | |||
AF336884.1 | - | 1 | 222 | |||
AF336886.1 | - | 1 | 150 | |||
AF336887.1 | - | 1 | 207 | |||
AF338190.1 | - | 1 | 195 | |||
AF338193.1 | - | 1 | 213 | |||
AF338195.1 | - | 1 | 213 | |||
AF338196.1 | - | 1 | 225 | |||
AF338198.1 | - | 1 | 213 | |||
AF338199.1 | - | 1 | 180 | |||
AF469044.1 | - | 1 | 162 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 22 | 2859 | ||
GenBank mRNA | BC063885.1 | - | 22 | 2845 | 30383658 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
