siRNA Details
Gene Symbol: SLC8A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033262.4 | NP_150287.1 | 2 | 1063 | ||
NM_058240.3 | NP_489479.1 | 2 | 1063 | |||
NM_182932.2 | NP_891977.1 | 2 | 1063 | |||
NM_183002.2 | NP_892114.1 | 2 | 1063 | |||
NR_104122.1 | 2 | 1063 | ||||
XM_017021606.1 | XP_016877095.1 | 2 | 435 | |||
XM_017021607.1 | XP_016877096.1 | 2 | 928 | |||
XM_017021608.1 | XP_016877097.1 | 2 | 689 | |||
XM_017021609.1 | XP_016877098.1 | 2 | 928 | |||
XM_017021610.1 | XP_016877099.1 | 2 | 928 | |||
GenBank mRNA | AF510501.1 | - | 2 | 1063 | ||
AF510502.1 | - | 2 | 1063 | |||
AF510503.1 | - | 2 | 1063 | |||
AJ304852.1 | - | 1 | 371 | |||
AJ304853.1 | - | 1 | 371 | |||
BC036783.1 | - | 2 | 866 | |||
BC150183.1 | - | 1 | 386 | |||
BC150193.1 | - | 1 | 386 | |||
BI906774.1 | - | 2 | 519 | |||
BX248763.1 | - | 2 | 927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
