siRNA Details
Gene Symbol: SPATA31A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015667.2 | NP_056482.2 | 4 | 2079 | ||
GenBank mRNA | BC040528.1 | - | 4 | 2079 | 4838470 |
Gene Symbol: SPATA31A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083124.1 | NP_001076593.1 | 4 | 2079 | ||
XM_011545680.2 | XP_011543982.1 | 3 | 1996 | |||
GenBank mRNA | AL080148.1 | - | 1 | 1034 |
Gene Symbol: SPATA31A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113541.1 | NP_001107013.1 | 4 | 2079 | ||
XM_011545671.2 | XP_011543973.1 | 4 | 2150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
