siRNA Details
Gene Symbol: ARL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113738.1 | NP_001107210.1 | 1, 2 | 128 | ||
NM_001288811.1 | NP_001275740.1 | 1, 2 | 128 | |||
NM_001288812.1 | NP_001275741.1 | 1, 2 | 128 | |||
NM_001288813.1 | NP_001275742.1 | 1, 2 | 128 | |||
NM_016632.2 | NP_057716.2 | 1, 2 | 128 | |||
XM_005257439.4 | XP_005257496.1 | 1, 2 | 187 | |||
XM_011524875.2 | XP_011523177.1 | 1, 2 | 181 | |||
XM_011524876.2 | XP_011523178.1 | 1, 2 | 180 | |||
XM_011524877.2 | XP_011523179.1 | 1, 2 | 176 | |||
XM_011524878.2 | XP_011523180.1 | 1, 2 | 178 | |||
XM_017024729.1 | XP_016880218.1 | 1, 2 | 180 | |||
GenBank mRNA | AB209344.1 | - | 1, 2 | 59 | ||
AF208864.1 | - | 1, 2 | 76 | |||
AK289590.1 | - | 1, 2 | 66 | |||
AK315857.1 | - | 1, 2 | 67 | |||
BC020869.1 | - | 1, 2 | 49 | |||
BC030570.1 | - | 1, 2 | 99 | 5518676 | ||
BC041803.1 | - | 1, 2 | 85 | 5303659 | ||
BX647266.1 | - | 1, 2 | 88 | |||
DA761795.1 | - | 1, 2 | 77 | |||
DA979569.1 | - | 1, 2 | 59 | |||
DB027627.1 | - | 1, 2 | 88 | |||
DB065209.1 | - | 1, 2 | 128 | |||
DB241677.1 | - | 1, 2 | 108 |
Gene Symbol: ARL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039083.3 | NP_001034172.3 | 1, 2 | 99 | ||
NM_001103154.1 | NP_001096624.1 | 1, 2 | 99 | |||
XM_005256918.4 | XP_005256975.1 | 1, 2 | 203 | |||
XM_011524160.2 | XP_011522462.1 | 1, 2 | 199 | |||
XM_011524161.2 | XP_011522463.1 | 1, 2 | 200 | |||
XM_011524162.2 | XP_011522464.1 | 1, 2 | 200 | |||
XM_011524163.2 | XP_011522465.1 | 1, 2 | 200 | |||
XM_011524164.2 | XP_011522466.1 | 1, 2 | 196 | |||
XM_011524165.2 | XP_011522467.1 | 1, 2 | 201 | |||
XM_011524166.2 | XP_011522468.1 | 1, 2 | 199 | |||
XM_011524167.2 | XP_011522469.1 | 1, 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
