siRNA Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894991.1 | - | 1 | 13 | ||
BC029444.1 | - | 1 | 39 | |||
BC056256.1 | - | 1 | 49 | |||
BC062732.1 | - | 1 | 44 | |||
BC067092.1 | - | 1 | 36 | |||
BC067226.1 | - | 1 | 47 | |||
BC070334.1 | - | 1 | 44 | |||
BC073764.1 | - | 1 | 30 | |||
BC073791.1 | - | 1 | 35 | |||
BC095489.1 | - | 1 | 39 | |||
BC110292.1 | - | 1 | 27 |
Gene Symbol: IGKV1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC034146.1 | - | 1 | 44 |
Gene Symbol: IGKV1D-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U86790.1 | - | 1 | 19 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
