siRNA Details
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033664.1 | 3 | 316 | |||
GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
4 | 436 | |
AY312368.1 | - | LIT3328 AAA1 |
3 | 316 | ||
AY312370.1 | - | LIT3333 AAA1 |
3 | 201 | ||
AY312371.1 | - | LIT3334 AAA1 |
3 | 316 | ||
AY321516.1 | - | 3 | 197 | |||
BC130477.1 | - | 3 | 201 | |||
BX649082.1 | - | 3 | 279 | |||
U85653.1 | - | 4 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
