siRNA Details
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033664.1 | 4, 5 | 531 | |||
NR_033665.1 | 3, 4 | 381 | ||||
GenBank mRNA | AY312368.1 | - | LIT3328 AAA1 |
4, 5 | 531 | |
AY312369.1 | - | LIT3332 AAA1 |
3, 4 | 381 | ||
AY312370.1 | - | LIT3333 AAA1 |
4, 5 | 416 | ||
AY321516.1 | - | 4, 5 | 412 | |||
BX649082.1 | - | 4, 5 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
