siRNA Details
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033664.1 | 5 | 601 | |||
NR_033665.1 | 4 | 451 | ||||
GenBank mRNA | AY312368.1 | - | LIT3328 AAA1 |
5 | 601 | |
AY312369.1 | - | LIT3332 AAA1 |
4 | 451 | ||
AY312370.1 | - | LIT3333 AAA1 |
5 | 486 | ||
AY321516.1 | - | 5 | 482 | |||
BX649082.1 | - | 5 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
