siRNA Details
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 11 | 954 | ||
XM_011521988.2 | XP_011520290.1 | 11 | 1639 | |||
XM_011521989.2 | XP_011520291.1 | 11 | 1643 | |||
GenBank mRNA | BQ477899.1 | - | 6 | 484 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 10 | 1256 | |||
GenBank mRNA | AK292534.1 | - | 10 | 1139 | ||
BC110417.1 | - | 10 | 1227 | 5171794 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 11 | 952 |
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 10 | 1155 | |||
GenBank mRNA | AK093104.1 | - | 10 | 1155 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 11 | 954 | ||
XM_006720651.3 | XP_006720714.1 | 11 | 974 | |||
XM_011521948.2 | XP_011520250.1 | 11 | 1282 | |||
XM_011521950.2 | XP_011520252.1 | 11 | 1280 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 11 | 954 |
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 11 | 954 | ||
XM_005268284.4 | XP_005268341.1 | 11 | 1753 | |||
XM_005268285.4 | XP_005268342.1 | 11 | 1753 | |||
XM_006720654.3 | XP_006720717.1 | 11 | 897 | |||
XM_011521951.2 | XP_011520253.1 | 12 | 1322 | |||
XM_011521952.2 | XP_011520254.1 | 11 | 1753 | |||
XM_011521953.2 | XP_011520255.1 | 11 | 1753 | |||
XM_011521954.2 | XP_011520256.1 | 11 | 887 | |||
GenBank mRNA | BF087405.1 | - | 3 | 226 |
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 11 | 954 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 11 | 951 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 10 | 1230 |
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 11 | 974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
