siRNA Details
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 11, 12 | 1526 | |||
GenBank mRNA | AK302625.1 | - | 12, 13 | 1227 | ||
BC144672.1 | - | 9, 10 | 1223 | |||
BC150302.1 | - | 11, 12 | 1500 | 8860399 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 11, 12 | 1526 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 11, 12 | 1526 | |||
GenBank mRNA | AK292534.1 | - | 11, 12 | 1409 | ||
BC110417.1 | - | 11, 12 | 1497 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 11, 12 | 1526 | |||
GenBank mRNA | BC063309.1 | - | 11, 12 | 1500 | ||
DQ309037.1 | - | 11, 12 | 1500 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 11, 12 | 1526 | |||
GenBank mRNA | BC066774.1 | - | 11, 12 | 1500 | ||
BC099913.1 | - | 10, 11 | 2353 | 4838463 | ||
BC125193.1 | - | 10, 11 | 1318 | 40122746 | ||
BC125194.1 | - | 10, 11 | 1318 | 40122749 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 11, 12 | 1479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
