siRNA Details
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 16, 17 | 1578 | ||
XM_005268284.4 | XP_005268341.1 | 16, 17 | 2377 | |||
XM_005268285.4 | XP_005268342.1 | 15, 16 | 2308 | |||
XM_006720654.3 | XP_006720717.1 | 16, 17 | 1521 | |||
XM_011521951.2 | XP_011520253.1 | 17, 18 | 1946 | |||
XM_011521954.2 | XP_011520256.1 | 16, 17 | 1511 |
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 16, 17 | 1578 |
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 16, 17 | 1578 | ||
XM_011521988.2 | XP_011520290.1 | 15, 16 | 2194 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 16, 17 | 1575 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 15, 16 | 1845 |
Gene Symbol: LOC101930434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022809.1 | XP_016878298.1 | 5, 6 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
