siRNA Details
Gene Symbol: MAFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002359.3 | NP_002350.1 | 3 | 3801 | ||
NM_032711.3 | NP_116100.2 | 3 | 3833 | |||
XM_011523578.1 | XP_011521880.1 | 3 | 4736 | |||
GenBank mRNA | AK130699.1 | - | 1 | 774 | ||
BC006143.1 | - | 1 | 184 | |||
BC103722.1 | - | 1 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
