siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 2 | 291 | ||
NM_002629.3 | NP_002620.1 | 2 | 361 | |||
GenBank mRNA | AK223096.1 | - | 2 | 207 | ||
AK292216.1 | - | 2 | 262 | |||
AK293023.1 | - | 2 | 206 | |||
AK295991.1 | - | 2 | 213 | |||
AK296619.1 | - | 2 | 150 | |||
AK312351.1 | - | 2 | 190 | |||
AK315910.1 | - | 2 | 294 | |||
AY007118.1 | - | 1 | 198 | |||
BC000455.1 | - | 1 | 224 | 2821567 | ||
BC010038.1 | - | 1 | 216 | 3604026 | ||
BC011678.2 | - | 1 | 180 | 4299008 | ||
BC053356.1 | - | 1 | 197 | 5787678 | ||
BC062302.1 | - | 1 | 196 | 6584466 | ||
BC066959.1 | - | 2 | 229 | 5262935 | ||
BC073742.1 | - | 1 | 195 | 6297173 | ||
BC157873.1 | - | 2 | 213 | 8862699 | ||
CR541659.1 | - | 2 | 178 | |||
CR541744.1 | - | 2 | 178 | |||
DA270129.1 | - | 2 | 361 | |||
EU794644.1 | - | 2 | 190 | |||
J04173.1 | - | 2 | 209 |
Gene Symbol: PGAM1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC171813.1 | - | 1 | 232 | 9054467 |
Gene Symbol: PGAM1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077225.1 | 5 | 630 | |||
GenBank mRNA | BC022928.1 | - | 5 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
