siRNA Details
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300981.1 | NP_001287910.1 | 3, 4 | 346 | ||
NM_021130.4 | NP_066953.1 | 2, 3 | 189 | |||
GenBank mRNA | AB451307.1 | - | 2, 3 | 100 | ||
AB451438.1 | - | 2, 3 | 100 | |||
AK026569.1 | - | 1 | 144 | |||
AK130101.1 | - | 2, 3 | 145 | |||
AK290085.1 | - | 2, 3 | 145 | |||
AK290851.1 | - | 2, 3 | 147 | |||
AK293003.1 | - | 3, 4 | 302 | |||
AK297340.1 | - | 2, 3 | 144 | |||
AK316564.1 | - | 2, 3 | 144 | |||
AU100120.1 | - | 2, 3 | 183 | |||
BC000689.2 | - | 1 | 122 | 3349335 | ||
BC003026.1 | - | 1 | 128 | 2823490 | ||
BC005320.1 | - | 1 | 144 | 3935025 | ||
BC005982.1 | - | 1 | 147 | 4109260 | ||
BC007104.1 | - | 1 | 145 | 4281376 | ||
BC013915.1 | - | 1 | 149 | 3684721 | ||
BC018843.2 | - | 1 | 179 | |||
BC073992.1 | - | 1 | 134 | 6161389 | ||
BC093076.1 | - | 3, 4 | 211 | 30414589 | ||
BC106030.1 | - | 1 | 136 | 2986147 | ||
BC137057.1 | - | 2, 3 | 189 | 9021054 | ||
BC137058.1 | - | 2, 3 | 189 | 9021055 | ||
BM806224.1 | - | 3, 4 | 200 | |||
BU786475.1 | - | 2, 3 | 183 | |||
CD014106.1 | - | 3, 4 | 302 | |||
CR456707.1 | - | 2, 3 | 100 | |||
EU794650.1 | - | 1 | 136 | |||
Y00052.1 | - | 2, 3 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
